Learn More
Aviva Systems Biology Corp CCR4 Antibody - middle region (AVARP07017_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP07017_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCR4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCR4. Alternate names: CC-CKR-4, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5, MGC88293, CD194ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-62544