Learn More
Eurofins Discoverx Ready-to-Assay CCR4 Chemokine Receptor Frozen Cells, 2 Vials of Cells. (HTS009RTA)
Catalog No. 502005572
Encompass
The Ready-to-Assay CCR4 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. MIP, RANTES) based activation of CCR4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCR4 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CCR4, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5572 Supplier Eurofins Discoverx Supplier No. HTS009RTA
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Ready-to-Assay CCR4 Chemokine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. MIP, RANTES) based activation of CCR4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses CCR4 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of CCR4, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.