Learn More
Aviva Systems Biology Corp CCR5 Antibody - middle region (ARP30788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCR5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCR5. Alternate names: CC-CKR-5, CCCKR5, CD195, CKR-5, CKR5, CMKBR5, IDDM22ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-54167