Learn More
Medchemexpress LLC HY-112701 10mg Medchemexpress, CCR6 inhibitor 1 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11270110MG
Encompass_Preferred
Medchemexpress, HY-112701 10mg CCR6 inhibitor 1 CAS: CCR6 inhibitor 1 is a potent and selective CCR6 inhibitor, with IC50s of 0.45 and 6 nM for monkey and human CCR6, much more selective at CCR6 over human CCR1 (IC50, > 30000 nM), and CCR7 (IC50, 9400 nM). CCR6 inhibitor 1 markedly blocks ERK phosphorylation. CCR6 inhibitor 1 is used in the research of autoimmune diseases and cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9145