Learn More
TARGETMOL CHEMICALS INC CCR6 INHIBITOR 1 5MG
SDP
Catalog No. 502581686
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCR6 inhibitor 1 is an effective and selective inhibitor of CCR6 (IC50 of monkey and human CCR6 was 0.45 nM and 6 nM respectively). CCR6 is associated with both autoimmune and non-autoimmune diseases so CCR6 inhibitor 1 is useful for in vitro and in vivo pathophysiology studies as a small molecule inhibitor of CCR6. purity: 98%
Catalog No. 50-258-1686 Supplier TARGETMOL CHEMICALS INC Supplier No. T107155MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCR6 inhibitor 1 is an effective and selective inhibitor of CCR6 (IC50 of monkey and human CCR6 was 0.45 nM and 6 nM respectively). CCR6 is associated with both autoimmune and non-autoimmune diseases so CCR6 inhibitor 1 is useful for in vitro and in vivo pathophysiology studies as a small molecule inhibitor of CCR6. purity: 98%