Learn More
Enzo Life Sciences CCR7 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ALX210844C100
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 2-22 (D2PGKPRKNVLVVALLVIFQVC22) of mouse CCR7 (C-C chemokine receptor 7). Species reactivity: Human, Mouse. Applications: Flow Cytometry, ICC, IHC (PS), WB. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide.
Catalog No. 50-200-8172