Learn More
Aviva Systems Biology Corp CCR8 Antibody - middle region (AVARP09059_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP09059_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CCR8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCR8. Alternate names: CY6, TER1, CCR-8, CKRL1, CDw198, CMKBR8, GPRCY6, CMKBRL2, CC-CKR-8ApplicationWB. Species reactivity: Cow, Human, Rabbit
Catalog No. 50-014-62595