Learn More
Biomatik Corporation Ccr8 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07071
Encompass_Preferred
Ccr8 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to CCR8. The Ccr8 Recombinant Antibody has been validated for the following applications: FC. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: Recombinant Human Ccr8 Protein. Applications: FC. Target Synonyms: CCR8. Accession Number: P51685. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7792