Learn More
Enzo Life Sciences CCR8 (mouse) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ALX210845C200
Encompass_Preferred
Host: Goat. Immunogen: Synthetic peptide corresponding to aa 7-33 (P7NVTMTDYYPDFFTAPCDAEFLLRGSM33) of mouse CCR8 (C-C chemokine receptor 8). UniProt ID: P56484. Species reactivity: Mouse. Applications: ICC, IHC (PS), WB. Recommended Dilutions: Immunocytochemistry (1:500) Immunohistochemistry (paraffin sections (1:400)) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.1% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8173