Learn More
Biomatik Corporation Ccr9 Recombinant Antibody
Supplier:  Biomatik Corporation CAC10227
Encompass_Preferred
Ccr9 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to CCR9. The Ccr9 Recombinant Antibody has been validated for the following applications: ELISA, WB, IF, IP. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: A Synthesized Peptide Derived From Human Ccr9. Applications: ELISA, WB, IF, IP. Target Synonyms: CCR9. Accession Number: P51686. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7398