Learn More
ATCC CCRF-CEM; Leukemia; Human (Homo sapiens)
Catalog No. 502382549
Encompass
CCRF-CEM [CCRF CEM] are human T lymphoblasts isolated from the peripheral blood of a female, Caucasian 4-year-old with acute lymphoblastic leukemia (ALL). This cell line is used for immuno-oncology and immunology research.
Catalog No. 50-238-2549 Supplier ATCC Supplier No. CCL119
Add to Cart
Add to Cart
CCRF-CEM [CCRF CEM] are human T lymphoblasts isolated from the peripheral blood of a female, Caucasian 4-year-old with acute lymphoblastic leukemia (ALL). This cell line is used for immuno-oncology and immunology research.