Learn More
ATCC CCRF-SBB Lymphoblast Human (Homo sapiens)
Catalog No. 502384917
Encompass
CCRF-SB cells are B lymphoblasts isolated from peripheral blood derived from an acute lymphoblastic leukemia (ALL) patient. They can be used in immune system disorder and immunology research.
Catalog No. 50-238-4917 Supplier ATCC Supplier No. CCL120
CCRF-SB cells are B lymphoblasts isolated from peripheral blood derived from an acute lymphoblastic leukemia (ALL) patient. They can be used in immune system disorder and immunology research.