Learn More
Aviva Systems Biology Corp CCRL1 Antibody - C-terminal region (ARP62100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCRL1. It was validated on Western Blot. . Alternate names: CC-CKR-11, CCBP2, CCR10, CCR11, CCX-CKR, CKR-11, PPR1, VSHK1, CCR-11, CCX CKR, CCRL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-568