Learn More
Aviva Systems Biology Corp CCRL2 Antibody - N-terminal region (ARP34023_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34023_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCRL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCRL2. Alternate names: CKRX, CRAM-A, CRAM-B, HCR, MGC116710, CRAMApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55222