Learn More
Aviva Systems Biology Corp NOCT Antibody - N-terminal region (ARP32536_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32536_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CCRN4L. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CCRN4L. Alternate names: NOC, CCR4LApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54640