Learn More
Aviva Systems Biology Corp CCS Antibody - middle region (ARP61008_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61008_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCS. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CCS. Alternate names: MGC138260ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-231