Learn More
TARGETMOL CHEMICALS INC CCT 031374 HYDROBROMIDE 5MG
SDP
Catalog No. 502581012
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCT 031374 hydrobromide is a TCF/(beta)-catenin inhibitor with antitumor activities. purity: 98%
Catalog No. 50-258-1012 Supplier TARGETMOL CHEMICALS INC Supplier No. T216855MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCT 031374 hydrobromide is a TCF/(beta)-catenin inhibitor with antitumor activities. purity: 98%