Learn More
TARGETMOL CHEMICALS INC CCT 137690 25MG
SDP
Catalog No. 502580718
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCT 137690 is a highly specific and oral-available aurora kinase inhibitor for aurora A(IC50=15 nM ) B(IC50=25 nM) and C(IC50=19 nM). purity: 99%
Catalog No. 50-258-0718 Supplier TARGETMOL CHEMICALS INC Supplier No. T261125MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CCT 137690 is a highly specific and oral-available aurora kinase inhibitor for aurora A(IC50=15 nM ) B(IC50=25 nM) and C(IC50=19 nM). purity: 99%