Learn More
Abcam CCT-251921, 1MG
Catalog No. 502671208
Encompass_Preferred
MW 410.9 Da, Purity >=98%. CCT-251921 is a potent, selective, small-molecule modulator of the mediator complex-associated kinases CDK8 and CDK19 (IC₅₀, CDK8 = 2.3 nM; CDK19 = 2.6 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1208 Supplier Abcam Supplier No. AB2870701MG
MW 410.9 Da, Purity >=98%. CCT-251921 is a potent, selective, small-molecule modulator of the mediator complex-associated kinases CDK8 and CDK19 (IC₅₀, CDK8 = 2.3 nM; CDK19 = 2.6 nM).
The product is subject to the following: Abcam Restricted Use Statement