Learn More
Adam Equipment CCT 32UH - Cruiser CCT Bench Counting Scales - Capacity: 70lb / 32kg, Readability: 0.00044lb / 0.2g
Catalog No. NC3964898
Encompass_Preferred
Ideal for shipping and receiving, warehouse use, and production facilities, the CCT offers intelligent functions that are practical for any counting application. With the CCT, it's easy to check the number of pieces in a carton or container, helping reduce waste when filling orders or packaging shipments. Intuitive software is user-friendly and keeps track of the count, automatically optimizing the unit weight for greater accuracy. The CCT features a compact design, making it easy to transport between locations within a facility during inventory procedures.
Catalog No. NC3964898 Supplier Adam Equipment Supplier No. CCT 32UH
Ideal for shipping and receiving, warehouse use, and production facilities, the CCT offers intelligent functions that are practical for any counting application. With the CCT, it's easy to check the number of pieces in a carton or container, helping reduce waste when filling orders or packaging shipments. Intuitive software is user-friendly and keeps track of the count, automatically optimizing the unit weight for greater accuracy. The CCT features a compact design, making it easy to transport between locations within a facility during inventory procedures.