Learn More
Medchemexpress LLC HY-119240 10mg Medchemexpress, CCT020312 CAS:324759-76-4 Purity:>98%
SDP
Catalog No. 501969520
Encompass_Preferred
Medchemexpress, HY-119240 10mg CCT020312 CAS:324759-76-4 CCT020312 is a selective EIF2AK3/PERK activator. CCT020312 elicits EIF2A phosphorylation in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9520 Supplier Medchemexpress LLC Supplier No. HY11924010MG
Medchemexpress, HY-119240 10mg CCT020312 CAS:324759-76-4 CCT020312 is a selective EIF2AK3/PERK activator. CCT020312 elicits EIF2A phosphorylation in cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.