Learn More
Selleck Chemical LLC CCT020312-E1030-5MG
SDP
Catalog No. 502390019
Encompass_Preferred
CCT020312 is a selective PERK (EIF2AK3) activator with an EC50 of 5 1 M CCT020312 can be used for the selective activation of EIF2A-mediated translation control
Catalog No. 50-239-0019 Supplier Selleck Chemical LLC Supplier No. E10305MG
CCT020312 is a selective PERK (EIF2AK3) activator with an EC50 of 5 1 M CCT020312 can be used for the selective activation of EIF2A-mediated translation control