Learn More
Medchemexpress LLC CCT031374 hydrobromide | 1219184-91-4 | 98.1% | 434.33 | 10 MG
SDP
Catalog No. 5000346850
Encompass_Preferred
CCT031374 hydrobromide is a potent inhibitor of β-catenin/transcription factor (TCF) complex signaling. It inhibits TCF-dependent transcription of genes of the Wnt signaling pathway and exhibits antitumor activity.
  • Potent inhibitor of β-catenin/TCF complex signaling
  • Inhibits TCF-dependent transcription of Wnt signaling pathway genes
  • Has antitumor activity
Catalog No. 50-003-46850 Supplier Medchemexpress LLC Supplier No. HY10844110MG
CCT031374 hydrobromide is a potent inhibitor of β-catenin/transcription factor (TCF) complex signaling. It inhibits TCF-dependent transcription of genes of the Wnt signaling pathway and exhibits antitumor activity.
  • Potent inhibitor of β-catenin/TCF complex signaling
  • Inhibits TCF-dependent transcription of Wnt signaling pathway genes
  • Has antitumor activity