Learn More
Medchemexpress LLC CCT031374 hydrobromide | 1219184-91-4 | 98.1% | 434.33 | 100 MG
SDP
Catalog No. 5000347133
Encompass_Preferred
CCT 031374 hydrobromide is a potent inhibitor of β-catenin/transcription factor (TCF) complex signaling. It inhibits TCF-dependent transcription of genes in the Wnt signaling pathway and exhibits antitumor activity.
  • Potent inhibitor of β-catenin/transcription factor (TCF) complex signaling.
  • Inhibits TCF-dependent transcription of genes of Wnt signaling pathway.
  • Has antitumor activity.
  • For research use only.
Catalog No. 50-003-47133 Supplier Medchemexpress LLC Supplier No. HY10844150MG
CCT 031374 hydrobromide is a potent inhibitor of β-catenin/transcription factor (TCF) complex signaling. It inhibits TCF-dependent transcription of genes in the Wnt signaling pathway and exhibits antitumor activity.
  • Potent inhibitor of β-catenin/transcription factor (TCF) complex signaling.
  • Inhibits TCF-dependent transcription of genes of Wnt signaling pathway.
  • Has antitumor activity.
  • For research use only.