Learn More
Cayman Chemical CCT031374hydrobromIde 10mg
SDP
Supplier:  Cayman Chemical 3286610 MG
Encompass_Preferred
An inhibitor of β-catenin/TCF complex signaling; inhibits TCF-dependent transcription induced by expression of ∆N-β-catenin but not expression of a constitutively active TCF-VP16 fusion protein in reporter assays at 30 µM; induces apoptosis in, and inhibits growth of, HCT116 colon cancer cells
Catalog No. 50-294-6155