Learn More
Enzo Life Sciences CCT036477 (5 mg). CAS: 305372-78-5
SDP
Supplier:  Enzo Life Sciences BMLWN1140005
Encompass_Preferred
Inhibits Wnt signaling via blocking transcription at the β-catenin level independent of CBP or p300. Inhibited growth in several cancer cell lines (HT29, HCT116, SW480, and SNU475). CCT036447 was also active in vivo where it blocked development of zebrafish and Xenopus embryos and the expression of Wnt target genes. Alternative name: N-((4-Chlorophenyl)(2-methyl-1H-indol-3-yl)methyl)pyridin-2-amine. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2369