Learn More
Abcam CCT128930, 5MG
Catalog No. 502682927
Encompass_Preferred
MW 341.8 Da, Purity =98%. Potent and selective AKT2 inhibitor (IC50= 6 nM) with selectivity greater than 28-fold and 20-fold over PKA kinase (IC50= 168 nM) and p70S6K (IC50= 120 nM), respectively.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2927 Supplier Abcam Supplier No. AB2735335MG
MW 341.8 Da, Purity =98%. Potent and selective AKT2 inhibitor (IC50= 6 nM) with selectivity greater than 28-fold and 20-fold over PKA kinase (IC50= 168 nM) and p70S6K (IC50= 120 nM), respectively.
The product is subject to the following: Abcam Restricted Use Statement