Learn More
Apexbio Technology LLC CCT128930 HCl, 5mg. Cas: 885499-61-6 (free base) MFCD: MFCD13177916. AKT inhibitor
SDP
Supplier:  Apexbio Technology LLC A86175
Encompass_Preferred
CCT128930 is a potent, selective, novel and ATP-competitive inhibitor for AKT2 (IC50= 6 nM). It also has 28-fold and 20-fold selectivity over PKA kinase (IC50= 168 nM) and p70S6K (IC50= 120 nM), respectively. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5165