Learn More
Apexbio Technology LLC CCT128930 HCl 885499-61-6 (free base) 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061791
Encompass_Preferred
CCT128930 HCl is a potent selective ATP-competitive inhibitor targeting AKT2 kinase with an IC50 of approximately 6 nM It exhibits higher selectivity for AKT2 compared to related kinases such as PKA and p70S6K Akt protein kinase involved in diverse cellular pathways including metabolism proliferation apoptosis transcription control and cell mobility is frequently deregulated in various tumor types In cancer cell models CCT128930 HCl impairs cellular proliferation and reduces phosphorylation of specific Akt downstream substrates Preclinical studies employing PTEN-deficient glioma cells and breast cancer xenograft models show inhibition of signaling activity and tumor growth Consequently CCT128930 HCl serves as a promising research tool for studying Akt-mediated signaling mechanisms pathway regulation and potential therapeutic responses in cancer biology
Catalog No. 50-000-61791 Supplier Apexbio Technology LLC Supplier No. A86175.1
CCT128930 HCl is a potent selective ATP-competitive inhibitor targeting AKT2 kinase with an IC50 of approximately 6 nM It exhibits higher selectivity for AKT2 compared to related kinases such as PKA and p70S6K Akt protein kinase involved in diverse cellular pathways including metabolism proliferation apoptosis transcription control and cell mobility is frequently deregulated in various tumor types In cancer cell models CCT128930 HCl impairs cellular proliferation and reduces phosphorylation of specific Akt downstream substrates Preclinical studies employing PTEN-deficient glioma cells and breast cancer xenograft models show inhibition of signaling activity and tumor growth Consequently CCT128930 HCl serves as a promising research tool for studying Akt-mediated signaling mechanisms pathway regulation and potential therapeutic responses in cancer biology