Learn More
Medchemexpress LLC CCT128930 hydrochloride | 2453324-32-6 | 99.0% | C18H21Cl2N5 | 10MG
SDP
Catalog No. 5000258326
Encompass_Preferred
CCT128930 hydrochloride is the hydrochloride salt of a selective ATP-competitive AKT inhibitor supplied as a powder for research use. It is employed in cellular assays to probe PI3K/Akt/mTOR signaling, induce cell cycle arrest, and study autophagy; handle and store according to the product SDS and COA.
Catalog No. 50-002-58326 Supplier Medchemexpress LLC Supplier No. HY13260A10MG
CCT128930 hydrochloride is the hydrochloride salt of a selective ATP-competitive AKT inhibitor supplied as a powder for research use. It is employed in cellular assays to probe PI3K/Akt/mTOR signaling, induce cell cycle arrest, and study autophagy; handle and store according to the product SDS and COA.