Learn More
TARGETMOL CHEMICALS INC CCT129202 5MG
SDP
Catalog No. 502579375
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCT129202 is an ATP-competitive pan-Aurora inhibitor for Aurora A Aurora B and Aurora C with IC50 of 0.042 uM 0.198 uM and 0.227 uM respectively. It is less potent to FGFR3 GSK3(beta) PDGFR(beta) etc. purity: 98%
Catalog No. 50-257-9375 Supplier TARGETMOL CHEMICALS INC Supplier No. T64355MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CCT129202 is an ATP-competitive pan-Aurora inhibitor for Aurora A Aurora B and Aurora C with IC50 of 0.042 uM 0.198 uM and 0.227 uM respectively. It is less potent to FGFR3 GSK3(beta) PDGFR(beta) etc. purity: 98%