Learn More
Medchemexpress LLC HY-12846 5mg Medchemexpress, CCT196969 CAS:1163719-56-9 Purity:>98%
SDP
Catalog No. 502081169
Encompass_Preferred
Medchemexpress, HY-12846 5mg CCT196969 CAS:1163719-56-9 CCT196969 is a pan-Raf inhibitor, which inhibits B-Raf, BRafV600E and CRAF with IC50s of 0.1, 0.04, and 0.01 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1169 Supplier Medchemexpress LLC Supplier No. HY128465MG
Medchemexpress, HY-12846 5mg CCT196969 CAS:1163719-56-9 CCT196969 is a pan-Raf inhibitor, which inhibits B-Raf, BRafV600E and CRAF with IC50s of 0.1, 0.04, and 0.01 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.