Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCT2 Human His-SUM 10ug
SDP

Catalog No. 5000195470
Encompass_Preferred

Recombinant human CCT2 supplied lyophilized with N-terminal 6xHis and SUMO tags, expressed in E. coli and intended for research use. The product is provided as a 10 μg vial, formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), and characterized for purity, molecular weight, endotoxin, and storage stability to support biochemical and cell biology applications.

  • Recombinant human CCT2 with N-terminal His and SUMO tags.
  • Expressed in E. coli for scalable production.
  • Lyophilized formulation in Tris-EDTA-trehalose buffer for stability.
  • Greater than 90% purity by reducing SDS-PAGE.
  • Molecular weight approximately 66 kDa.
  • Endotoxin level below 1 EU/μg by LAL assay.
  • Provided as a 10 μg vial; reconstitute to ≥100 μg/mL recommended.
  • Store at -20°C; after reconstitution stable short term at 4°C or long term at -20°C/-80°C.

Catalog No. 50-001-95470 Supplier Medchemexpress LLC Supplier No. HYP7212010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human CCT2 supplied lyophilized with N-terminal 6xHis and SUMO tags, expressed in E. coli and intended for research use. The product is provided as a 10 μg vial, formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), and characterized for purity, molecular weight, endotoxin, and storage stability to support biochemical and cell biology applications.

  • Recombinant human CCT2 with N-terminal His and SUMO tags.
  • Expressed in E. coli for scalable production.
  • Lyophilized formulation in Tris-EDTA-trehalose buffer for stability.
  • Greater than 90% purity by reducing SDS-PAGE.
  • Molecular weight approximately 66 kDa.
  • Endotoxin level below 1 EU/μg by LAL assay.
  • Provided as a 10 μg vial; reconstitute to ≥100 μg/mL recommended.
  • Store at -20°C; after reconstitution stable short term at 4°C or long term at -20°C/-80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.