Learn More
Medchemexpress LLC CCT2 Human His-SUM 5ug
SDP
Catalog No. 5000196262
Encompass_Preferred
Recombinant full-length human CCT2 (chaperonin containing TCP1 subunit 2) expressed in E. coli with N-terminal 6xHis and SUMO tags. Supplied as purified protein for biochemical, structural, and interaction studies; store frozen and aliquot to preserve integrity.
  • Purified full-length human CCT2 protein.
  • N-terminal 6xHis and SUMO tags facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~66 kDa for identification by SDS-PAGE.
  • Available in small microgram pack sizes suitable for assay development.
  • Stable when stored frozen; aliquot to minimize freeze-thaw cycles.
Catalog No. 50-001-96262 Supplier Medchemexpress LLC Supplier No. HYP721205UG
Recombinant full-length human CCT2 (chaperonin containing TCP1 subunit 2) expressed in E. coli with N-terminal 6xHis and SUMO tags. Supplied as purified protein for biochemical, structural, and interaction studies; store frozen and aliquot to preserve integrity.
  • Purified full-length human CCT2 protein.
  • N-terminal 6xHis and SUMO tags facilitate purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~66 kDa for identification by SDS-PAGE.
  • Available in small microgram pack sizes suitable for assay development.
  • Stable when stored frozen; aliquot to minimize freeze-thaw cycles.