Learn More
Medchemexpress LLC CCT241161 | 1163719-91-2 | 99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY128471MG
Encompass_Preferred
CCT241161 is an orally active pan-RAF inhibitor with IC50s of 3, 6, 10, 15 and 30 nM for LCK, CRAF, SRC, V600E-BRAF and BRAF, respectively. It demonstrates good activity in BRAF and NRAS mutant melanomas and also exhibits anticancer cell proliferative activity.
  • Orally active
  • Pan-RAF inhibitor
  • Shows activity in BRAF and NRAS mutant melanomas
  • Exhibits anticancer cell proliferative activity
Catalog No. 50-003-41831