Learn More
Medchemexpress LLC HY-14715B 5mg Medchemexpress, CCT241533 (hydrochloride) CAS:1431697-96-9 Purity:>98%
SDP
Catalog No. 502030552
Encompass_Preferred
Medchemexpress, HY-14715B 5mg CCT241533 (hydrochloride) CAS:1431697-96-9 CCT241533 hydrochloride is a potent and selective CHK2 inhibitor with an IC50 of 3 nM and a Ki of 1.16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0552 Supplier Medchemexpress LLC Supplier No. HY14715B5MG
Medchemexpress, HY-14715B 5mg CCT241533 (hydrochloride) CAS:1431697-96-9 CCT241533 hydrochloride is a potent and selective CHK2 inhibitor with an IC50 of 3 nM and a Ki of 1.16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.