Learn More
Cayman Chemical CCT241533 (hydrochloride), Purity: ≥98%, CAS: 1431697-96-9, 5 mg
SDP
Catalog No. 502982697
Encompass_Preferred
A selective Chk2 inhibitor (IC50 = 3 nM); potentiates the cytotoxicity of the PARP inhibitors, rucaparib and olaparib
Catalog No. 50-298-2697 Supplier Cayman Chemical Supplier No. 191785 MG
A selective Chk2 inhibitor (IC50 = 3 nM); potentiates the cytotoxicity of the PARP inhibitors, rucaparib and olaparib