Learn More
Medchemexpress LLC HY-18161 5mg Medchemexpress, CCT241736 CAS:1402709-93-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY181615MG
Encompass_Preferred
Medchemexpress, HY-18161 5mg CCT241736 CAS:1402709-93-6 CCT241736 is a potent and orally bioavailable dual FLT3 and Aurora kinase inhibitor, which inhibits Aurora kinases (Aurora-A Kd, 7.5 nM, IC50, 38 nM; Aurora-B Kd, 48 nM), FLT3 kinase (Kd, 6.2 nM), and FLT3 mutants including FLT3-ITD (Kd, 38 nM) and FLT3(D835Y) (Kd, 14 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2020