Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCT365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 | C18H18ClNO4S3 | 100MG
SDP

Catalog No. 5000237928
Encompass_Preferred

CCT365623 hydrochloride is a research-grade small molecule lysyl oxidase (LOX) inhibitor used in biochemical and cellular studies. It is supplied as a solid (white to yellow) with reported potency IC50 0.89 μM against LOX. Analytical purity is 98.07%, molecular weight 443.99, and formula C18H18ClNO4S3. The compound is highly soluble in DMSO (250 mg/mL) and is hygroscopic in solution; store sealed and away from moisture, with solvent storage recommended at -80°C (up to 6 months) or -20°C (up to 1 month).

  • Potent LOX inhibition with reported IC50 0.89 μM.
  • High analytical purity of 98.07%.
  • Molecular weight 443.99.
  • Chemical formula C18H18ClNO4S3.
  • Soluble in DMSO at 250 mg/mL; ultrasonic aid may be required.
  • Solid, white to yellow; hygroscopic in DMSO.
  • Store sealed away from moisture; solvent storage -80°C (6 months).

Catalog No. 50-002-37928 Supplier Medchemexpress LLC Supplier No. HY124674A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCT365623 hydrochloride is a research-grade small molecule lysyl oxidase (LOX) inhibitor used in biochemical and cellular studies. It is supplied as a solid (white to yellow) with reported potency IC50 0.89 μM against LOX. Analytical purity is 98.07%, molecular weight 443.99, and formula C18H18ClNO4S3. The compound is highly soluble in DMSO (250 mg/mL) and is hygroscopic in solution; store sealed and away from moisture, with solvent storage recommended at -80°C (up to 6 months) or -20°C (up to 1 month).

  • Potent LOX inhibition with reported IC50 0.89 μM.
  • High analytical purity of 98.07%.
  • Molecular weight 443.99.
  • Chemical formula C18H18ClNO4S3.
  • Soluble in DMSO at 250 mg/mL; ultrasonic aid may be required.
  • Solid, white to yellow; hygroscopic in DMSO.
  • Store sealed away from moisture; solvent storage -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.