Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cct365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 | C18H18ClNO4S3 | 10MG
SDP

Catalog No. 5000238817
Encompass_Preferred

CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor used for preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation, is orally active in models, and has demonstrated favorable tolerability and pharmacokinetic properties.

  • Orally active LOX inhibitor (IC50 = 0.89 μM).
  • High purity (98.1%) suitable for biological studies.
  • Available in small research pack sizes for in vitro and in vivo use.
  • Soluble in DMSO (250 mg/mL); in vivo formulation protocols provided.
  • Storage guidance included for solid and solution stability.
  • Intended for research use only; not for human administration.

Catalog No. 50-002-38817 Supplier Medchemexpress LLC Supplier No. HY124674A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor used for preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation, is orally active in models, and has demonstrated favorable tolerability and pharmacokinetic properties.

  • Orally active LOX inhibitor (IC50 = 0.89 μM).
  • High purity (98.1%) suitable for biological studies.
  • Available in small research pack sizes for in vitro and in vivo use.
  • Soluble in DMSO (250 mg/mL); in vivo formulation protocols provided.
  • Storage guidance included for solid and solution stability.
  • Intended for research use only; not for human administration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.