Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CCT365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 | C18H18ClNO4S3 | 10MM 1ML
SDP

Catalog No. 5000238109
Encompass_Preferred

CCT365623 hydrochloride is an orally active lysyl oxidase (LOX) inhibitor supplied as a high-purity research reagent. It is available as solids and as a 10 mM solution in DMSO for use in biochemical and cellular assays, with molecular weight 443.99 g/mol and chemical formula C18H18ClNO4S3. Typical applications include LOX inhibition studies, signaling pathway research, and preclinical investigations.

  • High purity, 98.1%.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5-100 mg).
  • Soluble in DMSO; ultrasonic agitation may improve dissolution.
  • Molecular weight 443.99 g/mol.
  • Suitable for biochemical and cellular LOX inhibition assays.
  • Provided with certificate of analysis for batch verification.
  • CAS number 2126136-98-7 for unambiguous identification.

Catalog No. 50-002-38109 Supplier Medchemexpress LLC Supplier No. HY124674A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CCT365623 hydrochloride is an orally active lysyl oxidase (LOX) inhibitor supplied as a high-purity research reagent. It is available as solids and as a 10 mM solution in DMSO for use in biochemical and cellular assays, with molecular weight 443.99 g/mol and chemical formula C18H18ClNO4S3. Typical applications include LOX inhibition studies, signaling pathway research, and preclinical investigations.

  • High purity, 98.1%.
  • Available as 10 mM solution in DMSO (1 mL) and as solids (5-100 mg).
  • Soluble in DMSO; ultrasonic agitation may improve dissolution.
  • Molecular weight 443.99 g/mol.
  • Suitable for biochemical and cellular LOX inhibition assays.
  • Provided with certificate of analysis for batch verification.
  • CAS number 2126136-98-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.