Learn More
Medchemexpress LLC CCT365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 | C18H18ClNO4S3 | 25MG
SDP
Catalog No. 5000237401
Encompass_Preferred
CCT365623 hydrochloride is an orally active lysyl oxidase (LOX) inhibitor intended for preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation driven by EGF, has an IC50 of 0.89 μM, and is reported to have good tolerability and pharmacokinetic properties.
  • Orally active lysyl oxidase inhibitor.
  • IC50 about 0.89 μM in biochemical assays.
  • Suppresses EGFR and AKT phosphorylation driven by EGF.
  • Reported good tolerability and pharmacokinetic properties in preclinical studies.
  • High chemical purity suitable for research applications.
  • Available as a 25 mg research package.
Catalog No. 50-002-37401 Supplier Medchemexpress LLC Supplier No. HY124674A25MG
CCT365623 hydrochloride is an orally active lysyl oxidase (LOX) inhibitor intended for preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation driven by EGF, has an IC50 of 0.89 μM, and is reported to have good tolerability and pharmacokinetic properties.
  • Orally active lysyl oxidase inhibitor.
  • IC50 about 0.89 μM in biochemical assays.
  • Suppresses EGFR and AKT phosphorylation driven by EGF.
  • Reported good tolerability and pharmacokinetic properties in preclinical studies.
  • High chemical purity suitable for research applications.
  • Available as a 25 mg research package.