Learn More
Medchemexpress LLC CCT365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 g·mol⁻¹ | C18H18ClNO4S3 | 50MG
SDP
Catalog No. 5000236417
Encompass_Preferred
CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor intended for research use. It exhibits LOX inhibitory activity in the submicromolar to low-micromolar range, suppresses EGF-driven EGFR (pY1068) and AKT phosphorylation in cellular studies, and has reported favorable pharmacokinetic properties.
  • Hydrochloride salt form for improved solubility.
  • Reported LOX inhibitory activity with IC50 ≈ 0.89 μM.
  • Suppresses EGF-driven EGFR (pY1068) and AKT phosphorylation in cell-based assays.
  • High reported purity (~98.1%).
  • Available as solid vials and as solution formulations in DMSO.
  • For research use only.
Catalog No. 50-002-36417 Supplier Medchemexpress LLC Supplier No. HY124674A50MG
CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor intended for research use. It exhibits LOX inhibitory activity in the submicromolar to low-micromolar range, suppresses EGF-driven EGFR (pY1068) and AKT phosphorylation in cellular studies, and has reported favorable pharmacokinetic properties.
  • Hydrochloride salt form for improved solubility.
  • Reported LOX inhibitory activity with IC50 ≈ 0.89 μM.
  • Suppresses EGF-driven EGFR (pY1068) and AKT phosphorylation in cell-based assays.
  • High reported purity (~98.1%).
  • Available as solid vials and as solution formulations in DMSO.
  • For research use only.