Learn More
Medchemexpress LLC CCT365623 hydrochloride | 2126136-98-7 | 98.1% | 443.99 g/mol | C18H18ClNO4S3 | 5MG
SDP
Catalog No. 5000237930
Encompass_Preferred
CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor used in preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation driven by EGF, and has reported oral activity and favorable pharmacokinetic properties. The material is supplied as a solid and as a prepared DMSO solution for laboratory use and is intended for research use only.
Catalog No. 50-002-37930 Supplier Medchemexpress LLC Supplier No. HY124674A5MG
CCT365623 hydrochloride is the hydrochloride salt of a small-molecule lysyl oxidase (LOX) inhibitor used in preclinical research. It suppresses EGFR (pY1068) and AKT phosphorylation driven by EGF, and has reported oral activity and favorable pharmacokinetic properties. The material is supplied as a solid and as a prepared DMSO solution for laboratory use and is intended for research use only.