Learn More
Medchemexpress LLC CCT367766 formic | 98.1% | 992.42 | 10 MG
SDP
Catalog No. 5000436268
Encompass_Preferred
CCT367766 formic is a potent, third-generation heterobifunctional and Cereblon-based pirin targeting protein degradation probe. It effectively depletes pirin protein expression at low concentrations and provides a potential chemical tool for studying this largely unexplored protein.
  • Exhibits moderate affinity for the CRBN-DDB1 complex with an IC50 of 490 nM.
  • Demonstrates good affinity for recombinant pirin and CRBN with Kd values of 55 nM and 120 nM, respectively.
  • Depletes pirin protein in SK-OV-3 human ovarian cancer cells in a time-dependent manner.
  • Rescues pirin expression from pretreatment of chlorobisamide in SK-OV-3 cells.
Catalog No. 50-004-36268 Supplier Medchemexpress LLC Supplier No. HY122653A10MG
CCT367766 formic is a potent, third-generation heterobifunctional and Cereblon-based pirin targeting protein degradation probe. It effectively depletes pirin protein expression at low concentrations and provides a potential chemical tool for studying this largely unexplored protein.
  • Exhibits moderate affinity for the CRBN-DDB1 complex with an IC50 of 490 nM.
  • Demonstrates good affinity for recombinant pirin and CRBN with Kd values of 55 nM and 120 nM, respectively.
  • Depletes pirin protein in SK-OV-3 human ovarian cancer cells in a time-dependent manner.
  • Rescues pirin expression from pretreatment of chlorobisamide in SK-OV-3 cells.