Learn More
Medchemexpress LLC CCT367766 formic | 98.1% | 992.42 g·mol-1 | C50H50ClN7O13 | 5 MG
SDP
Catalog No. 5000264381
Encompass_Preferred
CCT367766 formic is a heterobifunctional Cereblon-based PROTAC designed to induce targeted degradation of the pirin protein. It is intended for use as a chemical probe in mechanism-of-action studies and target validation, and is provided with analytical characterization data suitable for research applications.
  • Cereblon-based PROTAC that degrades pirin.
  • Third-generation heterobifunctional degrader for cellular target engagement studies.
  • High reported purity of 98.1% for reliable experimental use.
  • Characterized by LC-MS, HNMR, COA, and SDS documentation.
  • Reported binding affinities: pirin Kd ≈55 nM, CRBN Kd ≈120 nM, IC50 (CRBN-DDB1) ≈490 nM.
  • Available in small research quantities suitable for probe studies.
Catalog No. 50-002-64381 Supplier Medchemexpress LLC Supplier No. HY122653A5MG
CCT367766 formic is a heterobifunctional Cereblon-based PROTAC designed to induce targeted degradation of the pirin protein. It is intended for use as a chemical probe in mechanism-of-action studies and target validation, and is provided with analytical characterization data suitable for research applications.
  • Cereblon-based PROTAC that degrades pirin.
  • Third-generation heterobifunctional degrader for cellular target engagement studies.
  • High reported purity of 98.1% for reliable experimental use.
  • Characterized by LC-MS, HNMR, COA, and SDS documentation.
  • Reported binding affinities: pirin Kd ≈55 nM, CRBN Kd ≈120 nM, IC50 (CRBN-DDB1) ≈490 nM.
  • Available in small research quantities suitable for probe studies.