Learn More
Medchemexpress LLC HY-129188 5mg Medchemexpress, CCT369260 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1291885MG
Encompass_Preferred
Medchemexpress, HY-129188 5mg CCT369260 CAS: CCT369260 (compound 1) is an orally avtive B-cell lymphoma 6 (BCL6) inhibitor with anti-tumor activity. CCT369260 (compound 1) exhibits an IC50 of 520 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1199