Learn More
Aviva Systems Biology Corp CCT4 Antibody - C-terminal region (ARP34271_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34271_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CCT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CCT4. Alternate names: SRB, Cctd, CCT-DELTAApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55321