Learn More
Aviva Systems Biology Corp CCT5 Antibody - N-terminal region (ARP54825_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54825_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCT5. Alternate names: CCT-epsilon, CCTE, KIAA0098, TCP-1-epsilonApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61331