Learn More
ABclonal Technology CCT6B Rabbit pAb
SDP
Supplier:  ABclonal Technology A146151000UL
Encompass_Preferred
This gene encodes a molecular chaperone that is a member of the chaperonin-containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-06396